2-Cyanobenzenecarboximidamide hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Cyanobenzamidine hydrochloride

084788-1G
102843-376EA 750.77 USD
102843-376
2-Cyanobenzenecarboximidamide hydrochloride ≥97%
2-Cyanobenzenecarboximidamide hydrochloride

Hazard: Irritant

Formula: C₈H₈ClN₃
MW: 181.63 g/mol
MDL Number: MFCD16622254
CAS Number: 426828-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR