5-(2-Ethoxyphenyl)cyclohexane-1,3-dione ≥97%
Supplier: Matrix Scientific
084747-1G
102843-264EA
524.62
USD
102843-264
5-(2-Ethoxyphenyl)cyclohexane-1,3-dione ≥97%
5-(2-Ethoxyphenyl)cyclohexane-1,3-dione
Hazard: Irritant
Formula:
C₁₄H₁₆O₃ |
MDL Number:
MFCD11108768 CAS Number: 1092288-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...