5-Thien-3-ylcyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

084741-1G
102843-248EA 750.77 USD
102843-248
5-Thien-3-ylcyclohexane-1,3-dione ≥97%
5-Thien-3-ylcyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₀H₁₀O₂S
MW: 194.25 g/mol
MDL Number: MFCD12827893
CAS Number: 239132-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR