(2R)-2-(Benzyloxy)-1-(2,4-difluorophenyl)propan-1-one ≥97%

Supplier: Matrix Scientific

084740-1G
102843-246EA 949.23 USD
102843-246
(2R)-2-(Benzyloxy)-1-(2,4-difluorophenyl)propan-1-one ≥97%
(2R)-2-(Benzyloxy)-1-(2,4-difluorophenyl)propan-1-one

Hazard: Irritant

Formula: C₁₆H₁₄F₂O₂
MDL Number: MFCD12827892
CAS Number: 247196-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR