1-(3-Nitrophenyl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one ≥97%

Supplier: Matrix Scientific

084715-1G
102843-178EA 467.69 USD
102843-178
1-(3-Nitrophenyl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one ≥97%
1-(3-Nitrophenyl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one

Hazard: Irritant

Formula: C₁₆H₁₀F₃NO₃
MDL Number: MFCD04686949
CAS Number: 89807-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR