1-(3,5-Dichlorophenyl)-3-phenylprop-2-en-1-one ≥97%
Supplier: Matrix Scientific
084707-1G
102843-158EA
553.85
USD
102843-158
1-(3,5-Dichlorophenyl)-3-phenylprop-2-en-1-one ≥97%
1-(3,5-Dichlorophenyl)-3-phenylprop-2-en-1-one
Hazard: Irritant
Formula:
C₁₅H₁₀Cl₂O |
MDL Number:
MFCD12404901 CAS Number: 1217248-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...