3-(1,3-Benzodioxol-5-yl)-1-(3-nitrophenyl)prop-2-en-1-one ≥97%

Supplier: Matrix Scientific

084706-1G
102843-156EA 353.85 USD
102843-156
3-(1,3-Benzodioxol-5-yl)-1-(3-nitrophenyl)prop-2-en-1-one ≥97%
3-(1,3-Benzodioxol-5-yl)-1-(3-nitrophenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₆H₁₁NO₅
MDL Number: MFCD00115235
CAS Number: 1058712-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR