2-[(4-Nitrophenyl)thio]thiophene ≥97%

Supplier: Matrix Scientific

084699-1G
102843-140EA 524.62 USD
102843-140
2-[(4-Nitrophenyl)thio]thiophene ≥97%
2-[(4-Nitrophenyl)thio]thiophene

Hazard: Irritant

Formula: C₁₀H₇NO₂S₂
MDL Number: MFCD12404900
CAS Number: 857361-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR