2-(Pyrrolidin-1-ylsulfonyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084684-1G
102843-086EA 553.85 USD
102843-086
2-(Pyrrolidin-1-ylsulfonyl)benzonitrile ≥97%
2-(Pyrrolidin-1-ylsulfonyl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂S
MDL Number: MFCD03942313
CAS Number: 612044-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR