2-(Piperidin-1-ylsulfonyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084682-1G
102843-082EA 553.85 USD
102843-082
2-(Piperidin-1-ylsulfonyl)benzonitrile ≥97%
2-(Piperidin-1-ylsulfonyl)benzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂S
MDL Number: MFCD03998603
CAS Number: 440353-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR