2-(Morpholin-4-ylsulfonyl)benzonitrile ≥97%

Supplier: Matrix Scientific

084681-1G
102843-080EA 553.85 USD
102843-080
2-(Morpholin-4-ylsulfonyl)benzonitrile ≥97%
2-(Morpholin-4-ylsulfonyl)benzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD03998505
CAS Number: 612045-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR