2-Methyl-N-[(1E)-1-(2,4,6-trifluorophenyl)ethylidene]propane-2-sulfinamide ≥97%
Supplier: Matrix Scientific
084674-1G
102843-062EA
723.08
USD
102843-062
2-Methyl-N-[(1E)-1-(2,4,6-trifluorophenyl)ethylidene]propane-2-sulfinamide ≥97%
2-Methyl-N-[(1E)-1-(2,4,6-trifluorophenyl)ethylidene]propane-2-sulfinamide
Hazard: Irritant
Formula:
C₁₂H₁₄F₃NOS |
MDL Number:
MFCD26955537 CAS Number: 1356090-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...