N-(2-Fluoro-6-nitrophenyl)guanidine ≥97%

Supplier: Matrix Scientific

084667-1G
102843-040EA 723.08 USD
102843-040
N-(2-Fluoro-6-nitrophenyl)guanidine ≥97%
N-(2-Fluoro-6-nitrophenyl)guanidine

Hazard: Irritant

Formula: C₇H₇FN₄O₂
MDL Number: MFCD12026369
CAS Number: 1217015-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR