5-Pyridin-3-ylcyclohexane-1,3-dione ≥97%

Supplier: Matrix Scientific

084648-1G
102842-988EA 750.77 USD
102842-988
5-Pyridin-3-ylcyclohexane-1,3-dione ≥97%
5-Pyridin-3-ylcyclohexane-1,3-dione

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD11840301
CAS Number: 1216662-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR