1-Nitro-4-[(phenylsulfonyl)methyl]benzene ≥97%

Supplier: Matrix Scientific

084620-1G
102842-908EA 949.23 USD
102842-908
1-Nitro-4-[(phenylsulfonyl)methyl]benzene ≥97%
1-Nitro-4-[(phenylsulfonyl)methyl]benzene

Hazard: Irritant

Formula: C₁₃H₁₁NO₄S
MDL Number: MFCD00099030
CAS Number: 34063-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR