3-(2,4-Dimethylphenyl)-1H-pyrazole ≥97%

Supplier: Matrix Scientific

084605-1G
102842-868EA 353.85 USD
102842-868
3-(2,4-Dimethylphenyl)-1H-pyrazole ≥97%
3-(2,4-Dimethylphenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂
MDL Number: MFCD11505059
CAS Number: 154258-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR