(4-Piperazin-1-ylphenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

084514-1G
102842-622EA 383.08 USD
102842-622
(4-Piperazin-1-ylphenyl)acetonitrile ≥97%
(4-Piperazin-1-ylphenyl)acetonitrile

Hazard: Irritant

Formula: C₁₂H₁₅N₃
MDL Number: MFCD09924132
CAS Number: 868244-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR