(4-Hydrazinophenyl)acetonitrile ≥97%
Supplier: Matrix Scientific
084513-1G
102842-620EA
400
USD
102842-620
(4-Hydrazinophenyl)acetonitrile ≥97%
(4-Hydrazinophenyl)acetonitrile
Hazard: Irritant
Formula:
C₈H₉N₃ Melting Pt: 68…70 °C |
MDL Number:
MFCD09991818 CAS Number: 57411-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...