[5-(4,5-Dimethoxy-2-nitrophenyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid ≥97%

Supplier: Matrix Scientific

084509-1G
102842-608EA 553.85 USD
102842-608
[5-(4,5-Dimethoxy-2-nitrophenyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid ≥97%
[5-(4,5-Dimethoxy-2-nitrophenyl)-2-oxo-1,3,4-oxadiazol-3(2H)-yl]acetic acid

Hazard: Irritant

Formula: C₁₂H₁₁N₃O₈
MDL Number: MFCD09991816
CAS Number: 1216647-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR