[4-(4,4'-Bipiperidin-1-ylcarbonyl)phenyl]dimethylamine ≥97%

Supplier: Matrix Scientific

084501-1G
102842-592EA 496.92 USD
102842-592
[4-(4,4'-Bipiperidin-1-ylcarbonyl)phenyl]dimethylamine ≥97%
[4-(4,4'-Bipiperidin-1-ylcarbonyl)phenyl]dimethylamine

Hazard: Irritant

Formula: C₁₉H₂₉N₃O
MDL Number: MFCD09991811
CAS Number: 1216635-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR