N'-Phenylmethanesulfonohydrazide ≥97%

Supplier: Matrix Scientific

084466-1G
102842-478EA 283.08 USD
102842-478
N'-Phenylmethanesulfonohydrazide ≥97%
N'-Phenylmethanesulfonohydrazide

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
MDL Number: MFCD00631792
CAS Number: 109425-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR