1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

084445-1G
102842-430EA 806.15 USD
102842-430
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride ≥95%
1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride

Hazard: Irritant

Formula: C₈H₄ClF₃N₄O₂S
MDL Number: MFCD11505058
CAS Number: 1215564-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR