4-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}benzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

084417-1G
102842-354EA 806.15 USD
102842-354
4-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}benzenesulfonyl chloride ≥95%
4-{[2-(Trifluoromethyl)pyrimidin-4-yl]oxy}benzenesulfonyl chloride

Hazard: Irritant

Formula: C₁₁H₆ClF₃N₂O₃S
MDL Number: MFCD11100130
CAS Number: 1215542-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR