3-(Bromomethyl)-6,7-dichloroquinoxalin-2(1H)-one ≥95%

Supplier: Matrix Scientific

084395-1G
102842-292EA 1118.46 USD
102842-292
3-(Bromomethyl)-6,7-dichloroquinoxalin-2(1H)-one ≥95%
3-(Bromomethyl)-6,7-dichloroquinoxalin-2(1H)-one

Hazard: Irritant

Formula: C₉H₅BrCl₂N₂O
MDL Number: MFCD22566368
CAS Number: 1355334-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR