4-Chloro-2-(chloromethyl)thieno[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

084394-1G
102842-290EA 553.85 USD
102842-290
4-Chloro-2-(chloromethyl)thieno[2,3-d]pyrimidine ≥95%
4-Chloro-2-(chloromethyl)thieno[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂S
MDL Number: MFCD09991810
CAS Number: 147006-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR