6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

084393-1G
102842-288EA 949.23 USD
102842-288
6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine ≥95%
6-Bromo-2-(chloromethyl)-3-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₅BrClN₃O₂
MW: 290.51 g/mol
MDL Number: MFCD08704640
CAS Number: 708213-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR