1-(Chloromethyl)-3-methylenecyclobutanecarbonitrile ≥95%

Supplier: Matrix Scientific

084385-1G
102842-264EA 806.15 USD
102842-264
1-(Chloromethyl)-3-methylenecyclobutanecarbonitrile ≥95%
1-(Chloromethyl)-3-methylenecyclobutanecarbonitrile

Hazard: Irritant

Formula: C₇H₈ClN
MDL Number: MFCD10703548
CAS Number: 1086376-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR