4-Chloromethyl-5-methyl-2-(furan-3-yl)oxazole ≥95%

Supplier: Matrix Scientific

084357-1G
102842-190EA 636.92 USD
102842-190
4-Chloromethyl-5-methyl-2-(furan-3-yl)oxazole ≥95%
4-Chloromethyl-5-methyl-2-(furan-3-yl)oxazole

Hazard: Irritant

Formula: C₉H₈ClNO₂
MDL Number: MFCD08056338
CAS Number: 914637-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR