1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole ≥95%

Supplier: Matrix Scientific

084312-1G
102842-056EA 949.23 USD
102842-056
1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole ≥95%
1-Benzyl-4-isocyanato-5-methyl-1H-1,2,3-triazole

Hazard: Irritant

Formula: C₁₁H₁₀N₄O
MDL Number: MFCD08235261
CAS Number: 914637-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR