4-Bromo-2-(isocyanatomethyl)-1-methylbenzene ≥95%

Supplier: Matrix Scientific

084300-1G
102842-016EA 1767.69 USD
102842-016
4-Bromo-2-(isocyanatomethyl)-1-methylbenzene ≥95%
4-Bromo-2-(isocyanatomethyl)-1-methylbenzene

Hazard: Irritant

Formula: C₉H₈BrNO
MDL Number: MFCD09038676
CAS Number: 343314-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR