[1-(4-Chlorophenyl)cyclohexyl]methanol ≥97%
Supplier: Matrix Scientific
084283-1G
102841-974EA
1089.23
USD
102841-974
[1-(4-Chlorophenyl)cyclohexyl]methanol ≥97%
[1-(4-Chlorophenyl)cyclohexyl]methanol
Hazard: Irritant
Formula:
C₁₃H₁₇ClO |
MDL Number:
MFCD00019391 CAS Number: 95266-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...