(3-Methoxythien-2-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)-3-methoxythiophene

084220-1G
102841-824EA 313.85 USD
102841-824
(3-Methoxythien-2-yl)methanol ≥97%
(3-Methoxythien-2-yl)methanol

Hazard: Irritant

Formula: C₆H₈O₂S
MW: 144.19 g/mol
MDL Number: MFCD11846258
CAS Number: 220139-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR