(3{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)methanol ≥97%

Supplier: Matrix Scientific

084183-1G
102841-736EA 666.15 USD
102841-736
(3{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)methanol ≥97%
(3{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)methanol

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₂
MDL Number: MFCD11505053
CAS Number: 1215524-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR