3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide ≥97%

Supplier: Matrix Scientific

084146-1G
102841-628EA 750.77 USD
102841-628
3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide ≥97%
3-Chloroimidazo[1,2-a]pyridine-6-carbohydrazide

Hazard: Irritant

Formula: C₈H₇ClN₄O
MDL Number: MFCD11100124
CAS Number: 1211364-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR