8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide ≥97%

Supplier: Matrix Scientific

084139-1G
102841-610EA 849.23 USD
102841-610
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide ≥97%
8-Bromo-6-chloroimidazo[1,2-a]pyridine-2-carbohydrazide

Hazard: Irritant

Formula: C₈H₆BrClN₄O
MW: 289.53 g/mol
MDL Number: MFCD09951830
CAS Number: 1031927-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR