6-Bromo-8-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

084138-1G
102841-608EA 609.23 USD
102841-608
6-Bromo-8-nitroimidazo[1,2-a]pyridine ≥97%
6-Bromo-8-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD09836167
CAS Number: 1031927-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR