7-Chloro-2-methylimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

084129-1G
102841-582EA 636.92 USD
102841-582
7-Chloro-2-methylimidazo[1,2-a]pyridine ≥97%
7-Chloro-2-methylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇ClN₂
MDL Number: MFCD09991805
CAS Number: 1086376-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR