7-Bromo-1-methyl-5-nitro-1H-benzimidazole ≥97%
Supplier: Matrix Scientific
084096-1G
102841-510EA
1118.46
USD
102841-510
7-Bromo-1-methyl-5-nitro-1H-benzimidazole ≥97%
7-Bromo-1-methyl-5-nitro-1H-benzimidazole
Hazard: Irritant
Formula:
C₈H₆BrN₃O₂ |
MDL Number:
MFCD14666506 CAS Number: 1287217-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...