7-Bromo-5-nitro-1H-benzimidazole ≥97%

Supplier: Matrix Scientific

084094-1G
102841-506EA 863.08 USD
102841-506
7-Bromo-5-nitro-1H-benzimidazole ≥97%
7-Bromo-5-nitro-1H-benzimidazole

Hazard: Irritant

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD14666504
CAS Number: 206759-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR