4-(4-Chlorophenyl)-4-oxo-2-piperidin-1-ylbutanoic acid ≥97%

Supplier: Matrix Scientific

083065-1G
102838-830EA 1118.46 USD
102838-830
4-(4-Chlorophenyl)-4-oxo-2-piperidin-1-ylbutanoic acid ≥97%
4-(4-Chlorophenyl)-4-oxo-2-piperidin-1-ylbutanoic acid

Hazard: Irritant

Formula: C₁₅H₁₈ClNO₃
MW: 295.77 g/mol
MDL Number: MFCD00693818
CAS Number: 70596-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR