(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
083005-1G
102838-628EA
723.08
USD
102838-628
(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid ≥97%
(7-Bromoimidazo[1,2-a]pyridin-2-yl)acetic acid
Hazard: Irritant
Formula:
C₉H₇BrN₂O₂ MW: 255.08 g/mol |
MDL Number:
MFCD11840279 CAS Number: 1217061-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...