8-Aminoimidazo[1,2-a]pyridine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082997-1G
102838-612EA 723.08 USD
102838-612
8-Aminoimidazo[1,2-a]pyridine-2-carboxylic acid ≥97%
8-Aminoimidazo[1,2-a]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₇N₃O₂
MDL Number: MFCD11505024
CAS Number: 1216646-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR