2-Methylimidazo[2,1-b][1,3,4]thiadiazole-6-carboxylic acid ≥97%

Supplier: Matrix Scientific

082975-1G
102838-564EA 1118.46 USD
102838-564
2-Methylimidazo[2,1-b][1,3,4]thiadiazole-6-carboxylic acid ≥97%
2-Methylimidazo[2,1-b][1,3,4]thiadiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₆H₅N₃O₂S
MDL Number: MFCD09414726
CAS Number: 96356-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR