5-Methylimidazo[1,2-a]pyrazine-2-carboxylic acid ≥97%
Supplier: Matrix Scientific
082959-1G
102838-524EA
1515.38
USD
102838-524
5-Methylimidazo[1,2-a]pyrazine-2-carboxylic acid ≥97%
5-Methylimidazo[1,2-a]pyrazine-2-carboxylic acid
Hazard: Irritant
Formula:
C₈H₇N₃O₂ |
MDL Number:
MFCD08235232 CAS Number: 190381-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...