6-Iodoimidazo[1,2-b]pyridazine-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082920-1G
102838-430EA 949.23 USD
102838-430
6-Iodoimidazo[1,2-b]pyridazine-2-carboxylic acid ≥97%
6-Iodoimidazo[1,2-b]pyridazine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₄IN₃O₂
MDL Number: MFCD12026355
CAS Number: 1216380-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR