2-[(4-Fluorobenzyl)amino]cyclopentanecarboxylic acid ≥97%
Supplier: Matrix Scientific
082906-1G
102838-394EA
949.23
USD
102838-394
2-[(4-Fluorobenzyl)amino]cyclopentanecarboxylic acid ≥97%
2-[(4-Fluorobenzyl)amino]cyclopentanecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₆FNO₂ |
MDL Number:
MFCD19706036 CAS Number: 1355334-63-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...