2-(Benzylamino)cyclopentanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082903-1G
102838-388EA 949.23 USD
102838-388
2-(Benzylamino)cyclopentanecarboxylic acid ≥97%
2-(Benzylamino)cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD19706026
CAS Number: 1355334-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR