1-Benzoylcyclopropanecarboxylic acid ≥97%

Supplier: Matrix Scientific

082898-1G
102838-378EA 949.23 USD
102838-378
1-Benzoylcyclopropanecarboxylic acid ≥97%
1-Benzoylcyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MDL Number: MFCD16622244
CAS Number: 79172-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR