2-(N-(5-Methylisoxazol-3-yl)carbamoyl)cyclohexanecarboxylic acid ≥95%
Supplier: Matrix Scientific
082880-1G
102838-332EA
636.92
USD
102838-332
2-(N-(5-Methylisoxazol-3-yl)carbamoyl)cyclohexanecarboxylic acid ≥95%
2-(N-(5-Methylisoxazol-3-yl)carbamoyl)cyclohexanecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₄ |
MDL Number:
MFCD01243889 CAS Number: 313385-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...