1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1-[4-(Methylsulphonyl)phenyl]-5-oxo-3-pyrrolidinecarboxylic acid

082846-1G
102838-244EA 496.92 USD
102838-244
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid ≥97%
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃S
MW: 251.31 g/mol
MDL Number: MFCD08444946
CAS Number: 63674-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR